Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 3 protein, mouse (hFc-tagged) | 20UG
SDP

Catalog No. 5000177030
Encompass_Preferred

Recombinant heterodimeric integrin alpha V beta 3 protein derived from mouse, produced in HEK293 cells and fused to a C-terminal human Fc (hFc) tag. Supplied as a 20 μg research reagent, it is intended for receptor-ligand interaction studies, cell adhesion assays, and signaling investigations.

  • Recombinant heterodimeric integrin alpha V beta 3 derived from mouse
  • Expressed in HEK293 cells to support mammalian post-translational modifications
  • C-terminal human Fc (hFc) tag for purification and detection
  • Suitable for receptor-ligand binding, cell adhesion, and signaling assays
  • Molecular weight ranges reported for subunits: 110-135 kDa and 50-60 kDa
  • Supplied as a 20 μg unit for laboratory research use

Catalog No. 50-001-77030 Supplier Medchemexpress LLC Supplier No. HYP70076020UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant heterodimeric integrin alpha V beta 3 protein derived from mouse, produced in HEK293 cells and fused to a C-terminal human Fc (hFc) tag. Supplied as a 20 μg research reagent, it is intended for receptor-ligand interaction studies, cell adhesion assays, and signaling investigations.

  • Recombinant heterodimeric integrin alpha V beta 3 derived from mouse
  • Expressed in HEK293 cells to support mammalian post-translational modifications
  • C-terminal human Fc (hFc) tag for purification and detection
  • Suitable for receptor-ligand binding, cell adhesion, and signaling assays
  • Molecular weight ranges reported for subunits: 110-135 kDa and 50-60 kDa
  • Supplied as a 20 μg unit for laboratory research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.