Learn More
Medchemexpress LLC Integrin alpha V bet 50ug | 50ug
SDP
Catalog No. 5000187830
Encompass_Preferred
Integrin alpha V beta 6 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Human (HEK293 His) has molecular weight of 130 95 KDa respectively
Catalog No. 50-001-87830 Supplier Medchemexpress LLC Supplier No. HYP77721A50UG
Integrin alpha V beta 6 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Human (HEK293 His) has molecular weight of 130 95 KDa respectively