Learn More
Medchemexpress LLC Integrin alpha V bet 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7772450UG
Encompass_Preferred
Integrin alpha V beta 6 Protein part of the integrin alpha chain family is featured Integrin alpha V beta 6 Protein Rhesus Macaque (HEK293 His) is a recombinant protein dimer complex containing Rhesus Macaque-derived Integrin alpha V beta 6 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 6 Protein Rhesus Macaque (HEK293 His) has molecular weight of 90-140 kDa
Catalog No. 50-001-87944