Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 6 protein (human, biotinylated, HEK293, His-Avi) | 50UG
SDP

Catalog No. 5000177032
Encompass_Preferred

Integrin alpha V beta 6 protein is a recombinant human integrin heterodimer produced in HEK293 cells, biotinylated and engineered with His-Avi tags. Supplied lyophilized from PBS with 8% trehalose, the protein is intended for research use and requires cold storage; follow the manufacturer's datasheet for reconstitution and handling.

  • Recombinant human integrin αVβ6, biotinylated and His-Avi tagged.
  • Produced in HEK293 cells to preserve mammalian post-translational modifications.
  • Lyophilized formulation in PBS with 8% trehalose for stability.
  • Molecular weight approximately 90-150 kDa.
  • Store lyophilized at -20°C; reconstituted stability: 4°C for 1 week or -20°C for longer.
  • Intended for research applications and not for therapeutic or diagnostic use.

Catalog No. 50-001-77032 Supplier Medchemexpress LLC Supplier No. HYP70076250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin alpha V beta 6 protein is a recombinant human integrin heterodimer produced in HEK293 cells, biotinylated and engineered with His-Avi tags. Supplied lyophilized from PBS with 8% trehalose, the protein is intended for research use and requires cold storage; follow the manufacturer's datasheet for reconstitution and handling.

  • Recombinant human integrin αVβ6, biotinylated and His-Avi tagged.
  • Produced in HEK293 cells to preserve mammalian post-translational modifications.
  • Lyophilized formulation in PBS with 8% trehalose for stability.
  • Molecular weight approximately 90-150 kDa.
  • Store lyophilized at -20°C; reconstituted stability: 4°C for 1 week or -20°C for longer.
  • Intended for research applications and not for therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.