Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 3 protein, mouse (HEK293, His) | >95.0% | 50UG
SDP

Catalog No. 5000177250
Encompass_Preferred

Integrin alpha V beta 3 protein, mouse (HEK293, His) is a recombinant heterodimeric integrin complex produced in HEK293 cells and supplied with a C-terminal His tag. It is intended for research use to support studies of integrin-mediated cell adhesion, signaling, and ligand interactions.

  • Recombinant heterodimeric integrin complex (ITGAV:ITGB3).
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Purity greater than 95% as determined by reducing Tris-Bis PAGE.
  • Observed molecular weight 90-130 kDa.
  • Available in small-scale sizes suitable for biochemical assays (20 μg, 50 μg, 100 μg).
  • For research use only; not for therapeutic or diagnostic use.

Catalog No. 50-001-77250 Supplier Medchemexpress LLC Supplier No. HYP70076150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Integrin alpha V beta 3 protein, mouse (HEK293, His) is a recombinant heterodimeric integrin complex produced in HEK293 cells and supplied with a C-terminal His tag. It is intended for research use to support studies of integrin-mediated cell adhesion, signaling, and ligand interactions.

  • Recombinant heterodimeric integrin complex (ITGAV:ITGB3).
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for detection and purification.
  • Purity greater than 95% as determined by reducing Tris-Bis PAGE.
  • Observed molecular weight 90-130 kDa.
  • Available in small-scale sizes suitable for biochemical assays (20 μg, 50 μg, 100 μg).
  • For research use only; not for therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.