Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin alpha V beta 8 protein, mouse (HEK293, His) | >95.0% | 50UG
SDP

Catalog No. 5000178044
Encompass_Preferred

Recombinant mouse integrin αVβ8 expressed in HEK293 cells with a His tag, provided as a purified heterodimer for research use. The protein is intended for biochemical assays, receptor-ligand interaction studies, and cell adhesion experiments; handle and store according to the supplier datasheet and SDS.

  • Recombinant heterodimeric integrin αVβ8 from mouse source.
  • HEK293 expression system preserves mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight range 80-140 kDa reflecting α and β subunits.
  • Supplied in microgram quantities suitable for in-vitro research.
  • Datasheet, SDS, and COA provided for batch-specific details.

Catalog No. 50-001-78044 Supplier Medchemexpress LLC Supplier No. HYP70076350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse integrin αVβ8 expressed in HEK293 cells with a His tag, provided as a purified heterodimer for research use. The protein is intended for biochemical assays, receptor-ligand interaction studies, and cell adhesion experiments; handle and store according to the supplier datasheet and SDS.

  • Recombinant heterodimeric integrin αVβ8 from mouse source.
  • HEK293 expression system preserves mammalian post-translational modifications.
  • His tag facilitates purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight range 80-140 kDa reflecting α and β subunits.
  • Supplied in microgram quantities suitable for in-vitro research.
  • Datasheet, SDS, and COA provided for batch-specific details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.