Learn More
Medchemexpress LLC Integrin alpha V bet 5ug
SDP
Catalog No. 5000179615
Encompass_Preferred
Integrin V beta 3 proteins specifically the -V (ITGAV) subunit are multifunctional receptors for ligands such as vitronectin and fibronectin and recognize RGD sequences ITGAV ITGB3 binds fractalkine and acts as a coreceptor in CX3CR1-dependent signaling Integrin alpha V beta 3 Protein Human (HEK293 C-His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 3 protein expressed by HEK293 with C-10 His labeled tag Integrin alpha V beta 3 Protein Human (HEK293 C-His) has molecular weight of 90 31 110-150 kDa respectively
Catalog No. 50-001-79615 Supplier Medchemexpress LLC Supplier No. HYP73868A5UG
Integrin V beta 3 proteins specifically the -V (ITGAV) subunit are multifunctional receptors for ligands such as vitronectin and fibronectin and recognize RGD sequences ITGAV ITGB3 binds fractalkine and acts as a coreceptor in CX3CR1-dependent signaling Integrin alpha V beta 3 Protein Human (HEK293 C-His) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 3 protein expressed by HEK293 with C-10 His labeled tag Integrin alpha V beta 3 Protein Human (HEK293 C-His) has molecular weight of 90 31 110-150 kDa respectively