Learn More
Medchemexpress LLC Integrin alpha V bet 5ug
SDP
Supplier:  Medchemexpress LLC HYP738655UG
Encompass_Preferred
Integrin V beta 5 proteins specifically the -V (ITGAV) subunit are multifunctional receptors for ligands such as vitronectin and fibronectin and recognize RGD sequences ITGAV ITGB3 binds fractalkine and acts as a coreceptor in CX3CR1-dependent signaling Integrin alpha V beta 8 Protein Human (HEK293 His solution) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 8 protein expressed by HEK293 with C-His labeled tag Integrin alpha V beta 8 Protein Human (HEK293 His solution) has molecular weight of 187 95 kDa
Catalog No. 50-001-79136