Learn More
ACROBiosystems Integrin alpha V beta 1 100ug
Catalog No. 502534552
Encompass_Preferred
Rat ITGAV&ITGB1 Heterodimer Protein His Tag&Tag Free (IT1-R53W5) is expressed from CHO cells. It contains AA Phe 31 - Pro 988 & Gln 21 - Asp 729 (Accession # NP_001385621.1 & P49134-1).
Catalog No. 50-253-4552 Supplier ACROBiosystems Supplier No. IT1R53W5100UG
Rat ITGAV&ITGB1 Heterodimer Protein His Tag&Tag Free (IT1-R53W5) is expressed from CHO cells. It contains AA Phe 31 - Pro 988 & Gln 21 - Asp 729 (Accession # NP_001385621.1 & P49134-1).