Learn More
ACROBiosystems Integrin alpha V beta 1 100ug
Catalog No. 502534557
Encompass_Preferred
Cynomolgus Integrin alpha V beta 1 (ITGAV&ITGB1) Heterodimer Protein His Tag&Tag Free (IT1-C52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Gln 161 - Asp 868 (Accession # A0A2K5WCD3-1 & A0A7N9D0D7).
Catalog No. 50-253-4557 Supplier ACROBiosystems Supplier No. IT1C52W4100UG
Cynomolgus Integrin alpha V beta 1 (ITGAV&ITGB1) Heterodimer Protein His Tag&Tag Free (IT1-C52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Gln 161 - Asp 868 (Accession # A0A2K5WCD3-1 & A0A7N9D0D7).