Learn More
ACROBiosystems Integrin alpha V beta 3 100ug
Supplier:  ACROBiosystems IT3C52W3100UG
Encompass_Preferred
Cynomolgus Integrin alpha V beta 3 (ITGAV&ITGB3) Heterodimer Protein His Tag&Tag Free (IT3-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Gly 27 - Asp 718 (Accession # A0A2K5WCD3-1 & A0A2K5WYN9).
Catalog No. 50-253-4548