Learn More
ACROBiosystems Biotinylated Human ITGAV&ITGB3 Heterodimer Protein, His,Avitag™&Tag Free (IT3-H82W9) is expressed from human 293 cells (HEK293). (Accession # NP_002201.1 (ITGAV) & AAI27667.1 (ITGB3)).
Supplier:  ACROBiosystems IT3H82W9200UG
Encompass_Preferred
Integrin alpha-V/beta-3 (ITGAV:ITGB3) is a receptor for cytotactin, fibronectin, laminin, matrix metalloproteinase-2, osteopontin, osteomodulin, prothrombin, thrombospondin, vitronectin and von Willebrand factor. Integrins alpha-IIb/beta-3 and alpha-V/beta-3 recognize the sequence R-G-D in a wide array of ligands. Also, Integrin alpha-V/beta-3 acts as a receptor for herpes virus 8/HHV-8, coxsackievirus A9, Hantaan virus, cytomegalovirus/HHV-5, human metapneumovirus, human parechovirus 1 and west nile virus. Furthermore, in case of HIV-1 infection, the interaction with extracellular viral Tat protein seems to enhance angiogenesis in Kaposi's sarcoma lesions.
Catalog No. 50-223-0886