Learn More
ACROBiosystems Integrin alpha V beta 5 100ug
Catalog No. 502534546
Encompass_Preferred
Cynomolgus Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein His Tag&Tag Free (IT5-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Leu 25 - Asn 719 (Accession # A0A2K5WCD3-1 & A0A2K5UEB4).
Catalog No. 50-253-4546 Supplier ACROBiosystems Supplier No. IT5C52W3100UG
Cynomolgus Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein His Tag&Tag Free (IT5-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Leu 25 - Asn 719 (Accession # A0A2K5WCD3-1 & A0A2K5UEB4).