Learn More
ACROBiosystems Biotinylated Mouse Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein, His,Avitag™&Tag Free
Catalog No. 502331800
Encompass_Preferred
Biotinylated Mouse ITGAV&ITGB5 Heterodimer Protein, His,Avitag&Tag Free (IT5-M82W9) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 & Gly 24 - Asn 719 (Accession # P43406-1 & O70309-1).
Catalog No. 50-233-1800 Supplier ACROBiosystems Supplier No. IT5M82W9200UG
Biotinylated Mouse ITGAV&ITGB5 Heterodimer Protein, His,Avitag&Tag Free (IT5-M82W9) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 & Gly 24 - Asn 719 (Accession # P43406-1 & O70309-1).