Learn More
ACROBiosystems Cynomolgus ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-C52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 (ITGAV) & Glu 43 - Ser 681 (ITGB8) (Accession # A0A2K5WCD3-1 (ITGAV) & G7P0S0-1 (ITGB8)).
Catalog No. 502230737
Encompass_Preferred
Integrin alpha V beta 8 (ITGAV & ITGB8 or ITGAVB8) is expressed in yolk sac, placenta, brain perivascular astrocytes, Schwann cells, renal glomerular mesangial cells and pulmonary epithelial cells. Unlike other alpha V integrins, ITGAVB8 does not appear to assume different activation states, and the cytoplasmic tail does not connect to the cytoskeleton. It binds ligands containing an RGD motif, including vitronectin, fibrin and the latency associated peptide (LAP) of the latent TGF-beta complex. High affinity binding of alpha V beta 8 to LAP allows proteolytic cleavage by MT1-MMP, which releases active TGF-beta. This mechanism differs from that of alpha V beta 6, the other alpha V integrin which can activate TGF-beta from latency through non-proteolytic mechanisms. Downstream effects of TGF-beta activation include control of cell growth and associated vascularization.
This item is not returnable. View return policy
Catalog No. 50-223-0737 Supplier ACROBiosystems Supplier No. IT8C52W9100UG
Integrin alpha V beta 8 (ITGAV & ITGB8 or ITGAVB8) is expressed in yolk sac, placenta, brain perivascular astrocytes, Schwann cells, renal glomerular mesangial cells and pulmonary epithelial cells. Unlike other alpha V integrins, ITGAVB8 does not appear to assume different activation states, and the cytoplasmic tail does not connect to the cytoskeleton. It binds ligands containing an RGD motif, including vitronectin, fibrin and the latency associated peptide (LAP) of the latent TGF-beta complex. High affinity binding of alpha V beta 8 to LAP allows proteolytic cleavage by MT1-MMP, which releases active TGF-beta. This mechanism differs from that of alpha V beta 6, the other alpha V integrin which can activate TGF-beta from latency through non-proteolytic mechanisms. Downstream effects of TGF-beta activation include control of cell growth and associated vascularization.