Learn More
ACROBiosystems Biotinylated Mouse Integrin Alpha V Beta 8 Heterodimer Protein (His, Avitag & Tag Free) - 25 μg
Catalog No. 502370265
Encompass_Preferred
Biotinylated mouse integrin alpha V beta 8 heterodimer is a recombinant Avitag-biotinylated protein produced in HEK293 cells and supplied lyophilized in a 25 μg format. It is MALS-verified and >95% pure, and is intended for use in binding and receptor interaction assays, including studies involving latent TGF-β1.
  • Lyophilized 25 μg format for long-term storage.
  • Avitag enzymatic biotinylation for site-specific labeling.
  • Produced in HEK293 cells to preserve mammalian glycosylation.
  • Purity greater than 95% as determined by SDS-PAGE.
  • MALS-verified molecular mass and expected migration on SDS-PAGE.
  • Validated for binding to latent TGF-β1 with established linear ranges.
  • Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw cycles.
Catalog No. 50-237-0265 Supplier ACROBiosystems Supplier No. IT8M52W725UG
Biotinylated mouse integrin alpha V beta 8 heterodimer is a recombinant Avitag-biotinylated protein produced in HEK293 cells and supplied lyophilized in a 25 μg format. It is MALS-verified and >95% pure, and is intended for use in binding and receptor interaction assays, including studies involving latent TGF-β1.
  • Lyophilized 25 μg format for long-term storage.
  • Avitag enzymatic biotinylation for site-specific labeling.
  • Produced in HEK293 cells to preserve mammalian glycosylation.
  • Purity greater than 95% as determined by SDS-PAGE.
  • MALS-verified molecular mass and expected migration on SDS-PAGE.
  • Validated for binding to latent TGF-β1 with established linear ranges.
  • Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw cycles.