Learn More
Medchemexpress LLC Integrin alpha X bet 50ug
SDP
Supplier:  Medchemexpress LLC HYP7386450UG
Encompass_Preferred
Integrin -X/ -2 is a receptor for fibrinogen that recognizes GPR sequences It plays a key role in inflammatory responses particularly in monocyte adhesion and chemotaxis Integrin alpha X beta 2 Protein Human (HEK293 Flag His) is a recombinant protein dimer complex containing human-derived Integrin alpha X beta 2 protein expressed by HEK293 with C-His C-Flag labeled tag
Catalog No. 50-001-79815