Learn More
Medchemexpress LLC Integrin alpha X bet 5ug
SDP
Catalog No. 5000179013
Encompass_Preferred
Integrin -X/ -2 is a receptor for fibrinogen that recognizes GPR sequences It plays a key role in inflammatory responses particularly in monocyte adhesion and chemotaxis Integrin alpha X beta 2 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha X beta 2 protein expressed by HEK293 with C-His labeled tag Integrin alpha X beta 2 Protein Human (HEK293 His) has molecular weight of 130-150 kDa (ITGAX) 85-96 kDa (ITGB2) respectively
Catalog No. 50-001-79013 Supplier Medchemexpress LLC Supplier No. HYP738635UG
Integrin -X/ -2 is a receptor for fibrinogen that recognizes GPR sequences It plays a key role in inflammatory responses particularly in monocyte adhesion and chemotaxis Integrin alpha X beta 2 Protein Human (HEK293 His) is a recombinant protein dimer complex containing human-derived Integrin alpha X beta 2 protein expressed by HEK293 with C-His labeled tag Integrin alpha X beta 2 Protein Human (HEK293 His) has molecular weight of 130-150 kDa (ITGAX) 85-96 kDa (ITGB2) respectively