Learn More
Cell Signaling Technology Integrin β6 Rabbit Monoclonal Antibody, Highly Specific, 100 µL
SDP
Supplier:  Cell Signaling Technology 95153S
Encompass
Integrin β6 (E4M9P) XP Rabbit Monoclonal Antibody is a highly specific recombinant antibody developed for studying Integrin Beta6. Validated for Western Blotting, IHC, and Immunofluorescence, it plays a crucial role in cell adhesion and migration, making it essential for research in wound healing and tumorigenesis. This antibody is unique for its strong reactivity to human integrin β6, providing reliable results for various applications.
Catalog No. NC2323510