Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin beta 1 Antibody (YA337) | 10 UL
SDP

Catalog No. 5000397495
Encompass_Preferred

Integrin beta 1 Antibody (YA337) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Integrin beta 1.

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Targets Integrin beta 1
  • Applications: WB, IHC-P, FC
  • Reactivity: Human, Mouse

Catalog No. 50-003-97495 Supplier Medchemexpress LLC Supplier No. HYP8041810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin beta 1 Antibody (YA337) is a rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Integrin beta 1.

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Targets Integrin beta 1
  • Applications: WB, IHC-P, FC
  • Reactivity: Human, Mouse

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.