Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin beta 1 antibody (YA337) | Protein A affinity purified | Predicted band size: 88 kDa, Observed band size: 120-140 kDa | 50 UL
SDP

Catalog No. 5000398944
Encompass_Preferred

Integrin beta 1 antibody (YA337) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Integrin beta 1. Integrin beta 1 is a multifunctional cell surface receptor crucial for cell-extracellular matrix interactions, forming heterodimers with various alpha subunits to mediate processes like cell adhesion and angiogenesis. Different alpha/beta-1 complexes serve as receptors for various pathogens.

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Application: WB, IHC-P, FC
  • Conjugation: Non-conjugated
  • Clonality: Monoclonal
  • Isotype: IgG
  • Formulation: Supplied in 1*TBS (pH 7.4), 0.05% BSA and 40% Glycerol with 0.05% sodium azide as preservative
  • Appearance: Liquid
  • Shipping: With blue ice

Catalog No. 50-003-98944 Supplier Medchemexpress LLC Supplier No. HYP8041850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin beta 1 antibody (YA337) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Integrin beta 1. Integrin beta 1 is a multifunctional cell surface receptor crucial for cell-extracellular matrix interactions, forming heterodimers with various alpha subunits to mediate processes like cell adhesion and angiogenesis. Different alpha/beta-1 complexes serve as receptors for various pathogens.

  • Host: Rabbit
  • Reactivity: Human, mouse
  • Application: WB, IHC-P, FC
  • Conjugation: Non-conjugated
  • Clonality: Monoclonal
  • Isotype: IgG
  • Formulation: Supplied in 1*TBS (pH 7.4), 0.05% BSA and 40% Glycerol with 0.05% sodium azide as preservative
  • Appearance: Liquid
  • Shipping: With blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.