Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin beta-1/CD29 protein, human (recombinant, His-tagged) | >95.0% | 100UG
SDP

Catalog No. 5000176563
Encompass_Preferred

Recombinant human integrin beta-1 (CD29) extracellular domain produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized with high purity and formulated for biochemical and cell biology applications such as binding studies and cell adhesion assays.

  • Recombinant human extracellular domain with C-terminal His tag.
  • Produced in HEK293 mammalian expression system.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Sequence corresponds to UniProt P05556-1 (Q21-D728).
  • Lyophilized from buffered saline formulation for stability.
  • Recommended storage at -20°C; aliquot and store at -80°C for long term.

Catalog No. 50-001-76563 Supplier Medchemexpress LLC Supplier No. HYP70257100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human integrin beta-1 (CD29) extracellular domain produced in HEK293 cells with a C-terminal His tag. Supplied lyophilized with high purity and formulated for biochemical and cell biology applications such as binding studies and cell adhesion assays.

  • Recombinant human extracellular domain with C-terminal His tag.
  • Produced in HEK293 mammalian expression system.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Sequence corresponds to UniProt P05556-1 (Q21-D728).
  • Lyophilized from buffered saline formulation for stability.
  • Recommended storage at -20°C; aliquot and store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.