Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Integrin beta 4 Antibody (YA3280) | 50 UL
SDP

Catalog No. 5000443988
Encompass_Preferred

Integrin beta 4 Antibody (YA3280) is an unconjugated IgG antibody (clone number: YA3280) that specifically targets Integrin beta 4. This antibody is suitable for Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P) applications, showing reactivity in both human and mouse samples. It has an observed molecular weight of 210 kDa.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Conjugate: Non-conjugated
  • Applications: Western blot (WB), immunohistochemistry-paraffin (IHC-P)
  • Species reactivity: Human, mouse
  • Isotype: IgG
  • Synonyms: CD104, gp150, Integrin beta 4 subunit, ITG B4, ITGB 4, Itgb4
  • Purification method: Affinity chromatography
  • Storage conditions: Store at -20°C for 1 year. Avoid repeated freeze/thaw cycles. Shipped with blue ice.
  • Purity: ≥98.0%

Catalog No. 50-004-43988 Supplier Medchemexpress LLC Supplier No. HYP8353550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Integrin beta 4 Antibody (YA3280) is an unconjugated IgG antibody (clone number: YA3280) that specifically targets Integrin beta 4. This antibody is suitable for Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P) applications, showing reactivity in both human and mouse samples. It has an observed molecular weight of 210 kDa.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Conjugate: Non-conjugated
  • Applications: Western blot (WB), immunohistochemistry-paraffin (IHC-P)
  • Species reactivity: Human, mouse
  • Isotype: IgG
  • Synonyms: CD104, gp150, Integrin beta 4 subunit, ITG B4, ITGB 4, Itgb4
  • Purification method: Affinity chromatography
  • Storage conditions: Store at -20°C for 1 year. Avoid repeated freeze/thaw cycles. Shipped with blue ice.
  • Purity: ≥98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.