Learn More
CPC Scientific Ac-Gly-Cys-Gly-Tyr-Gly-Arg-Gly-Asp-Ser-Pro-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RGDP007A
Encompass
This is fibronectin derived, integrin-binding peptide. It may be used for PEG hydrogel preparation.SEQUENCE: Ac-Gly-Cys-Gly-Tyr-Gly-Arg-Gly-Asp-Ser-Pro-Gly-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Ac-GCGYGRGDSPG-NH2MOLECULAR FORMULA: C42H63N15O16SMOLECULAR WEIGHT: 1066.12STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [278792-07-7]SYNONYMS: RESEARCH AREA: CancerREFERENCES:Raeber, G. et al. Biophys. J. 89, 1374 (2005).SKU(s): RGDP-007A
Catalog No. 50-211-0384