Learn More
Cusabio Technology LLC integrin-linked kinase
Catalog No. 5000049404
Encompass_Preferred
The Human integrin-linked kinase 1 (ILK-1) ELISA Kit is designed for the quantitative detection of ILK-1 in serum plasma tissue homogenates and cell lysates using a sandwich ELISA method This kit detects ILK-1 with a sensitivity of 9 77 pg/mL and offers a broad detection range from 39 06 pg/mL to 2500 pg/mL ensuring accurate measurement across varying sample concentrations The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours providing efficient results for research applications Absorbance is measured at 450 nm to quantify ILK-1 levels making it suitable for studies involving cellular signaling pathways cancer research or cardiovascular diseases where precise ILK-1 quantification is critical The kit s robust performance and standardized protocol ensure reliability in both routine laboratory and specialized experimental settings
Catalog No. 50-000-49404 Supplier Cusabio Technology LLC Supplier No. CSBE10005H
The Human integrin-linked kinase 1 (ILK-1) ELISA Kit is designed for the quantitative detection of ILK-1 in serum plasma tissue homogenates and cell lysates using a sandwich ELISA method This kit detects ILK-1 with a sensitivity of 9 77 pg/mL and offers a broad detection range from 39 06 pg/mL to 2500 pg/mL ensuring accurate measurement across varying sample concentrations The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours providing efficient results for research applications Absorbance is measured at 450 nm to quantify ILK-1 levels making it suitable for studies involving cellular signaling pathways cancer research or cardiovascular diseases where precise ILK-1 quantification is critical The kit s robust performance and standardized protocol ensure reliability in both routine laboratory and specialized experimental settings