Learn More
ACROBiosystems Human Integrin Alpha 1 Beta 1 Heterodimer Protein, His Tag, Low Endotoxin, 100 μg
Catalog No. 501956617
Encompass_Preferred
Human Integrin alpha 1 beta 1 (ITGA1 and ITGB1) heterodimer protein acts as a receptor for laminin and collagen. It plays a critical role in supporting vascular endothelial growth factor signaling, endothelial cell migration, and tumor angiogenesis, while recognizing the proline-hydroxylated sequence G-F-P-G-E-R in collagen. This protein is expressed in human 293 cells and provided in a lyophilized powder form.
  • Greater than 95% purity determined by SDS-PAGE and greater than 90% purity determined by SEC-MALS
  • Low endotoxin level of less than 0.01 EU per μg
  • Calculated molecular weight of 130.0 kDa (ITGA1) and 83.7 kDa (ITGB1)
  • Formulated in 50 mM Tris, 0.2 M Arginine, 150 mM NaCl, pH 7.5 with trehalose
Catalog No. 50-195-6617 Supplier ACROBiosystems Supplier No. IT1H52WB100UG
Human Integrin alpha 1 beta 1 (ITGA1 and ITGB1) heterodimer protein acts as a receptor for laminin and collagen. It plays a critical role in supporting vascular endothelial growth factor signaling, endothelial cell migration, and tumor angiogenesis, while recognizing the proline-hydroxylated sequence G-F-P-G-E-R in collagen. This protein is expressed in human 293 cells and provided in a lyophilized powder form.
  • Greater than 95% purity determined by SDS-PAGE and greater than 90% purity determined by SEC-MALS
  • Low endotoxin level of less than 0.01 EU per μg
  • Calculated molecular weight of 130.0 kDa (ITGA1) and 83.7 kDa (ITGB1)
  • Formulated in 50 mM Tris, 0.2 M Arginine, 150 mM NaCl, pH 7.5 with trehalose