Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha 2 beta 1 (ITGA2&ITGB1) Heterodimer Protein, His Tag&Tag Free
Catalog No. 501957048
Encompass_Preferred
Recombinant Protein;100UG;Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W6) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Thr 1132 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # P17301-1 (ITGA2) & P05556-1 (ITGB1)).;Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA2 and ITGB1, has a calculated MW of 127.6 kDa (ITGA2) and 83.7 kDa (ITGB1). Subunit ITGA2 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 150-160 kDa (ITGA2) and 105-135 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52W6) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Human Collagen I at 2 μg/mL (100 μL/well) can bind Human ITGA2&ITGB1 Heterodimer
Catalog No. 50-195-7048 Supplier ACROBiosystems Supplier No. IT1H52W6100UG
Recombinant Protein;100UG;Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W6) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Thr 1132 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # P17301-1 (ITGA2) & P05556-1 (ITGB1)).;Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA2 and ITGB1, has a calculated MW of 127.6 kDa (ITGA2) and 83.7 kDa (ITGB1). Subunit ITGA2 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 150-160 kDa (ITGA2) and 105-135 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52W6) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Human Collagen I at 2 μg/mL (100 μL/well) can bind Human ITGA2&ITGB1 Heterodimer