Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha 3 beta 1 (ITGA3&ITGB1) Heterodimer Protein, His Tag&Tag Free
Catalog No. 501956484
Encompass_Preferred
Recombinant Protein;1MG;Human ITGA3&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52Wc) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Glu 991 (ITGA3) & Gln 21 - Asp 728 (ITGB1) (Accession # P26006-2 (ITGA3) & P05556-1 (ITGB1)).;Human ITGA3&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA3 and ITGB1, has a calculated MW of 133.3 kDa (ITGA3) and 83.7 kDa (ITGB1). Subunit ITGA3 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus.
Catalog No. 50-195-6484 Supplier ACROBiosystems Supplier No. IT1H52WC1MG
Recombinant Protein;1MG;Human ITGA3&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52Wc) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Glu 991 (ITGA3) & Gln 21 - Asp 728 (ITGB1) (Accession # P26006-2 (ITGA3) & P05556-1 (ITGB1)).;Human ITGA3&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA3 and ITGB1, has a calculated MW of 133.3 kDa (ITGA3) and 83.7 kDa (ITGB1). Subunit ITGA3 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus.