Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein;HEK293;100UG;IT1,Integrin alpha 6 beta 1,ITGA6B1,ITGA6&ITGB1,CD49f,VLA-6
Catalog No. 501658352
Encompass_Preferred
Recombinant Protein;Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein;100UG;Recombinant Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein (Human ITGA6 & ITGB1) Phe 24 - Gly 1012 (Integrin alpha 6, P23229-2) and Gln 21 - Asp 728 (Integrin beta 1, P05556-1) was co-expressed in human 293 cells (HEK293) at ACRObiosystems.;Human ITGA6 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA6 and ITGB1, has a calculated MW of 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1). Subunit ITGA6 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 24 (ITGA6) & Gln 21 (ITGB1). As a result of glycosylation, the protein migrates as 120-130 kDa under reducing (R) condition, and 100-125 kDa and 130-150 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8352 Supplier ACROBiosystems Supplier No. IT1H52W7100UG
Recombinant Protein;Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein;100UG;Recombinant Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein (Human ITGA6 & ITGB1) Phe 24 - Gly 1012 (Integrin alpha 6, P23229-2) and Gln 21 - Asp 728 (Integrin beta 1, P05556-1) was co-expressed in human 293 cells (HEK293) at ACRObiosystems.;Human ITGA6 & ITGB1 Heterodimer Protein, produced by co-expression of ITGA6 and ITGB1, has a calculated MW of 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1). Subunit ITGA6 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 24 (ITGA6) & Gln 21 (ITGB1). As a result of glycosylation, the protein migrates as 120-130 kDa under reducing (R) condition, and 100-125 kDa and 130-150 kDa under non-reducing (NR) condition (SDS-PAGE).