Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha E beta 7 (ITGAE&ITGB7) Heterodimer Protein, His Tag&Tag Free
Catalog No. 501957250
Encompass_Preferred
Recombinant Protein;1MG;Human ITGAE&ITGB7 Heterodimer Protein, His Tag&Tag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) & Glu 20 - His 723 (ITGB7) (Accession # P38570-1 (ITGAE) & P26010-1 (ITGB7)).;Human ITGAE&ITGB7 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAE and ITGB7, has a calculated MW of 128.6 kDa (ITGAE) and 81.8 kDa (ITGB7). Subunit ITGAE is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB7 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 135-150 kDa (ITGAE) and 100-115 kDa (ITGB7) respectively due to glycosylation.
Catalog No. 50-195-7250 Supplier ACROBiosystems Supplier No. IT7H52W71MG
Recombinant Protein;1MG;Human ITGAE&ITGB7 Heterodimer Protein, His Tag&Tag Free (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) & Glu 20 - His 723 (ITGB7) (Accession # P38570-1 (ITGAE) & P26010-1 (ITGB7)).;Human ITGAE&ITGB7 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAE and ITGB7, has a calculated MW of 128.6 kDa (ITGAE) and 81.8 kDa (ITGB7). Subunit ITGAE is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB7 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 135-150 kDa (ITGAE) and 100-115 kDa (ITGB7) respectively due to glycosylation.