Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha E beta 7 (ITGAE&ITGB7) Heterodimer Protein;HEK293;50UG;IT7,Integrin alpha E beta 7,ITGAE&ITGB7,HML-1,CD103
Supplier: ACROBiosystems IT7H52W750UG
Recombinant Protein;Human Integrin alpha E beta 7 (ITGAE&ITGB7) Heterodimer Protein;50UG;Human ITGAE&ITGB7, His Tag (IT7-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Ser 1124 (ITGAE) & Glu 20 - His 723 (ITGB7) (Accession # P38570-1 (ITGAE) & P26010-1 (ITGB7)).;Human ITGAE & ITGB7 Heterodimer Protein, produced by co-expression of ITGAE and ITGB7, has a calculated MW of 128.6 kDa (ITGAE) and 81.8 kDa (ITGB7). Subunit ITGAE is fused with a polyhistidine tag at the C-terminus and subunit ITGB7 contains no tag. The reducing (R) protein migrates as 135-150 kDa (ITGAE) and 105-115 kDa (ITGB7) respectively due to glycosylation.;Immobilized Human ITGAE&ITGB7, His Tag (Cat. No. IT7-H52W7) at 5 μg/mL (100 μL/well) can bind Human E-Cadherin, Fc Tag (Cat. No. ECD-H5250) with a linear range of 0.08-2.5 μg/mL (QC tested).