Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Integrin alpha V beta 1 (ITGAV&ITGB1) Heterodimer Protein, His,Avitag™&Tag Free
Catalog No. 501957152
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (IT1-H82W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gln 21 - Asp 728 (ITGB1) (Accession # NP_002201.1 (ITGAV) & NP_002202.2 (ITGB1)).;Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free, produced by co-expression of ITGAV and ITGB1, has a calculated MW of 114.7 kDa (ITGAV) and 83.7 kDa (ITGB1). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™) and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 135-150 kDa (ITGAV) and 100-115 kDa (ITGB1) respectively due to glycosylation.;Immobilized Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT1-H82W6) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-7152 Supplier ACROBiosystems Supplier No. IT1H82W6200UG
Recombinant Protein;200UG;Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (IT1-H82W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gln 21 - Asp 728 (ITGB1) (Accession # NP_002201.1 (ITGAV) & NP_002202.2 (ITGB1)).;Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free, produced by co-expression of ITGAV and ITGB1, has a calculated MW of 114.7 kDa (ITGAV) and 83.7 kDa (ITGB1). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™) and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 135-150 kDa (ITGAV) and 100-115 kDa (ITGB1) respectively due to glycosylation.;Immobilized Fibronectin at 5 μg/mL (100 μL/well) can bind Biotinylated Human ITGAV&ITGB1 Heterodimer Protein, His,Avitag&Tag Free (Cat. No. IT1-H82W6) with a linear range of 0.039-0.625 μg/mL (QC tested).