Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT5H52W51MG
Encompass_Preferred
Recombinant Protein;1MG;Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (IT5-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 24 - Asn 719 (ITGB5) (Accession # NP_002201.1 (ITGAV) & NP_002204.2 (ITGB5)).;Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB5, has a calculated MW of 112.9 kDa (ITGAV) and 81.9 kDa (ITGB5). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic tail and subunit ITGB5 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGAV) and 85-95 kDa (ITGB5) respectively due to glycosylation.;Immobilized Vitronectin at 2 μg/mL (100 μL/well) can bind Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT5-H52W5) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-195-7437