Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;HEK293;1MG;IT6,Integrin alpha V beta 6,ITGAV&ITGB6,Integrin,ITGAV,ITGA,ITGB6,ITGB,ITGAVB6
Catalog No. 501658926
Encompass_Preferred
Recombinant Protein;Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;1MG;Human ITGAV&ITGB6, His Tag (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 22 - Asn 707 (ITGB6) (Accession # NP_002201 (ITGAV)& AAI21179 (ITGB6)).;Human ITGAV & ITGB6 Heterodimer Protein, produced by co-expression of ITGAV and ITGB6, has a calculated MW of 113 kDa (ITGAV) and 79.6 kDa (ITGB6). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB6 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gly 22 (ITGB6). The reducing (R) protein migrates as 145 kDa (ITGAV) and 110-130 kDa (ITGB6) respectively due to glycosylation.;Immobilized Human ITGAV&ITGB6, His Tag (Cat. No. IT6-H52E1) at 2μg/mL (100 μL/well) can bind Human TGF-Beta 1 / TGFB1, His Tag & Avi Tag with a linear range of 2-39 ng/mL (QC tested).;Immobilized Human Fibro
Catalog No. 50-165-8926 Supplier ACROBiosystems Supplier No. IT6H52E11MG
Recombinant Protein;Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;1MG;Human ITGAV&ITGB6, His Tag (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 22 - Asn 707 (ITGB6) (Accession # NP_002201 (ITGAV)& AAI21179 (ITGB6)).;Human ITGAV & ITGB6 Heterodimer Protein, produced by co-expression of ITGAV and ITGB6, has a calculated MW of 113 kDa (ITGAV) and 79.6 kDa (ITGB6). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB6 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gly 22 (ITGB6). The reducing (R) protein migrates as 145 kDa (ITGAV) and 110-130 kDa (ITGB6) respectively due to glycosylation.;Immobilized Human ITGAV&ITGB6, His Tag (Cat. No. IT6-H52E1) at 2μg/mL (100 μL/well) can bind Human TGF-Beta 1 / TGFB1, His Tag & Avi Tag with a linear range of 2-39 ng/mL (QC tested).;Immobilized Human Fibro