Learn More
ACROBiosystems ACROBiosystems Mouse Integrin alpha V beta 8 (ITGAV&ITGB8) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT8M52W61MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-M52W6) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 988 (ITGAV) & Gly 22 - Leu 681 (ITGB8) (Accession # P43406-1 (ITGAV) & Q0VBD0-1 (ITGB8)).;Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB8, has a calculated MW of 112.5 kDa (ITGAV) and 77.5 kDa (ITGB8). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB8 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGAV) and 80-90 kDa (ITGB8) respectively due to glycosylation.;Immobilized Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-M52W6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human Fibronectin at 2 μg/mL (100 μL/well)
Catalog No. 50-195-6994