Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha V beta 8 (ITGAV&ITGB8) Heterodimer Protein, His Tag&Tag Free
Catalog No. 501957436
Encompass_Preferred
Recombinant Protein;1MG;Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Glu 43 - Arg 684 (ITGB8) (Accession # NP_002201.1 (ITGAV) & NP_002205.1 (ITGB8)).;Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB8, has a calculated MW of 112.9 kDa (ITGAV) and 76.5 kDa (ITGB8). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic tail and subunit ITGB8 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGAV) and 80-90 kDa (ITGB8) respectively due to glycosylation.;Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 1-63 ng/mL (QC tested).
Catalog No. 50-195-7436 Supplier ACROBiosystems Supplier No. IT8H52W41MG
Recombinant Protein;1MG;Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Glu 43 - Arg 684 (ITGB8) (Accession # NP_002201.1 (ITGAV) & NP_002205.1 (ITGB8)).;Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAV and ITGB8, has a calculated MW of 112.9 kDa (ITGAV) and 76.5 kDa (ITGB8). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic tail and subunit ITGB8 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGAV) and 80-90 kDa (ITGB8) respectively due to glycosylation.;Immobilized Human ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 1-63 ng/mL (QC tested).