Learn More
DENTSPLY SIRONA Integrity TempGrip Temporary C&B Cement Mix Tips Refill 50
Catalog No. NC1686694
Encompass_Preferred
Integrity TempGrip Temporary Crown & Bridge Cement is a zinc-oxide, noneugenol cement with an automix syringe delivery. It provides high compressive and flexural strength, along with smooth removal, as it stays in the crown when the provisional is removed.
Catalog No. NC1686694 Supplier DENTSPLY SIRONA Supplier No. 666460
Integrity TempGrip Temporary Crown & Bridge Cement is a zinc-oxide, noneugenol cement with an automix syringe delivery. It provides high compressive and flexural strength, along with smooth removal, as it stays in the crown when the provisional is removed.