Learn More
Cole-Parmer INTELLI-MIXER SHAKER, RM-2L
Supplier:  Cole-Parmer 5120220
Encompass
Intelli-Mixer IMIX-01 RM-2L Large Multifunctional Vortex Shaker, 1 to 99 rpm. Powered by V-Spin the Intelli-Mixer RM-2 is the latest in cutting-edge science. V-spin harnesses the energy of a stepper motor, allowing for a wide variety of mixing, vibrating, and rotating cycles. Featuring 15 Mix, 12 rocking, 4 blotting and 3 custom modes all in one versatile unit.Mfr # IMIX-01 Rotator, vortex mixer, blotter all in one34 modes of operationCold room safe
Catalog No. NC2092426