Learn More
Hach Company IntelliCAL PHC705 pH probe Kit
Catalog No. 5000207524 Shop All Hach Company Products
Encompass
IntelliCAL PHC705 Laboratory High Alkalinity Range RedRod Refillable Glass pH Electrode with Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately
Catalog No. 50-002-07524 Supplier Hach Company Supplier No. PHC705A01AP
IntelliCAL PHC705 Laboratory High Alkalinity Range RedRod Refillable Glass pH Electrode with Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately