Learn More
Hach Company IntelliCAL PHC725 pH probe Kit
Catalog No. 5000207856 Shop All Hach Company Products
Encompass
IntelliCAL PHC725 Laboratory Low Ionic Strength Media RedRod Refillable Glass pH Electrode with Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately
Catalog No. 50-002-07856 Supplier Hach Company Supplier No. PHC72501AP
IntelliCAL PHC725 Laboratory Low Ionic Strength Media RedRod Refillable Glass pH Electrode with Reagents PackThe RedRod sensor kit comes with calibration buffers reagents and maintenance solutions to provide an all-in-one easy to use reliable solution for your calibrations helping you ensure accurate pH readings with your HQD pH Meter The 500 mL bottles allows up to 6 months of pH electrode calibration when calibrating once per week or up to 25 calibrations at a very attractive price compared if purchased separately