Learn More
RWD LIFE SCIENCE INC Intelligent Optogenetics System 465nm, 0-100mW, 0-500Hz Adjustable/635nm, 0-60mW, 0-500Hz Adjustable
SDP
Catalog No. NC3404373
Encompass_Preferred
RWD new intelligent optogenetics system, on the basis of the last generation of integrated machines (laser and waveform generator integrated into one), has made another leap in the promotion. The new system is equipped with remote control function, one-click test conversion and three signal triggering modes to meet diversified experimental requirements. Using this instrument, it can reduce the interference of human factors to animal behavior experiment, and the unique test program and experiment record output view function are more conducive to experiment management.
Catalog No. NC3404373 Supplier RWD LIFE SCIENCE INC Supplier No. IOS465/635
RWD new intelligent optogenetics system, on the basis of the last generation of integrated machines (laser and waveform generator integrated into one), has made another leap in the promotion. The new system is equipped with remote control function, one-click test conversion and three signal triggering modes to meet diversified experimental requirements. Using this instrument, it can reduce the interference of human factors to animal behavior experiment, and the unique test program and experiment record output view function are more conducive to experiment management.