Learn More
Azenta US Inc INTELLIXCAP 96-CHANNEL EXTND
Catalog No. NC3002308
Encompass
  • Facilitates automated workflow integration
  • Designed for taller 96-format tubes
  • Compatible with IntelliXcap 24, 48, and 96 decapper/recapper systems
  • Enhances versatility and throughput of laboratory processes
  • Accommodates tubes with integral swab caps
  • Suitable for diverse biological samples and compound libraries
Catalog No. NC3002308 Supplier Azenta US Inc Supplier No. 468112
  • Facilitates automated workflow integration
  • Designed for taller 96-format tubes
  • Compatible with IntelliXcap 24, 48, and 96 decapper/recapper systems
  • Enhances versatility and throughput of laboratory processes
  • Accommodates tubes with integral swab caps
  • Suitable for diverse biological samples and compound libraries