Learn More
Percival Scientific Percival Scientific IntellusUltra Control System, Solid-State Controller, Dual Temperature Control, Ambient Monitoring
SDP
Catalog No. NC3441601
Encompass_Preferred
The Percival Scientific IntellusUltra Control System is a programmable controller designed for precise environmental management in research chambers. It features advanced programming styles, dual experiment protection, and extensive alarm capabilities. Its user-friendly interface, field-upgradable firmware, and robust data logging ensure accurate control of experimental conditions, making it ideal for scientific research.
Catalog No. NC3441601 Supplier Percival Scientific Supplier No. I66VL
Add to Cart
Add to Cart
Request a Quote
The Percival Scientific IntellusUltra Control System is a programmable controller designed for precise environmental management in research chambers. It features advanced programming styles, dual experiment protection, and extensive alarm capabilities. Its user-friendly interface, field-upgradable firmware, and robust data logging ensure accurate control of experimental conditions, making it ideal for scientific research.