Learn More
Research Products International Corp Autoradiography High Speed Intensifying Screens, 14 x 17 Inches, 2 per Package
SDP
Catalog No. 50212729
Encompass
RPI high speed intensifying screens with calcium tungstate phosphors produce the highest quality image detail at reduced exposure times. Screens include double sided tape on the inactive surface for permanent mounting in a cassette. Compatible with all blue based x-ray films.
Catalog No. 50-212-729 Supplier Research Products International Corp Supplier No. 431417
RPI high speed intensifying screens with calcium tungstate phosphors produce the highest quality image detail at reduced exposure times. Screens include double sided tape on the inactive surface for permanent mounting in a cassette. Compatible with all blue based x-ray films.