Learn More
Medchemexpress LLC HY-14339 5mg Medchemexpress, Intepirdine CAS:607742-69-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY143395MG
Encompass_Preferred
Medchemexpress, HY-14339 5mg Intepirdine CAS:607742-69-8 Intepirdine (SB742457) is a highly selective 5-HT6 receptor antagonist with pKi of 9.63; exhibits >100-fold selectivity over other receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1588