Learn More
Athens Research And Technology Inc Inter Alpha Inhibitor Proteins (IαIp), Native Human Plasma, ≥90% (SDS-PAGE), 25 µg
Catalog No. 50489826
Encompass_Preferred
Inter Alpha Inhibitor Proteins (IαIp) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied frozen in a 25 µg pack. Molecular weight: 225 kDa and 125 kDa. Inter Alpha Inhibitor Protein (IaIp), Human Plasma: 25 UG; Frozen in 1x PBS; MW 225,000 & 125,000; Purity: > =90% by SDS-PAGE. Molecular Biology Reagent
Catalog No. 50-489-826 Supplier Athens Research And Technology Inc Supplier No. 161609010925UG
Inter Alpha Inhibitor Proteins (IαIp) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied frozen in a 25 µg pack. Molecular weight: 225 kDa and 125 kDa. Inter Alpha Inhibitor Protein (IaIp), Human Plasma: 25 UG; Frozen in 1x PBS; MW 225,000 & 125,000; Purity: > =90% by SDS-PAGE. Molecular Biology Reagent