Learn More
Andwin Scientific INTER CRYOMOLD 1200/CS
SDP
Catalog No. NC9025044
Encompass_Preferred
Tissue-tek Intermediate Cryomold, Molds disposable Plastic cryomolds create a uniformly shaped flat-surface specimen block when used with O.C.T compound 25608-930 for frozen sectioning, 15L x 15W x 5mm dia., 1200/CS
Catalog No. NC9025044 Supplier Andwin Scientific Supplier No. 25608924CS
Tissue-tek Intermediate Cryomold, Molds disposable Plastic cryomolds create a uniformly shaped flat-surface specimen block when used with O.C.T compound 25608-930 for frozen sectioning, 15L x 15W x 5mm dia., 1200/CS