Learn More
MSC Interapid Contact Point, Carbide, 0.100 in Diameter, 2.75 in Length
Catalog No. 5000529172
Encompass
This contact point is made from extra carbide, providing durability and precision for various measuring applications. It features a 0.100 inch diameter and a 2.75 inch length, making it suitable for specific measurement needs.
  • Made from extra carbide.
  • Features a 0.100 inch diameter.
  • Offers a 2.75 inch length.
Catalog No. 50-005-29172 Supplier MSC Supplier No. 01746445
This contact point is made from extra carbide, providing durability and precision for various measuring applications. It features a 0.100 inch diameter and a 2.75 inch length, making it suitable for specific measurement needs.
  • Made from extra carbide.
  • Features a 0.100 inch diameter.
  • Offers a 2.75 inch length.