Learn More
MSC INTERAPID Test Indicator Contact Point, 0.0800 in Ball Dia, Carbide
Catalog No. 5000536534
Encompass
This test indicator contact point, with a 0.0800 in ball diameter, is crafted from durable carbide. It is engineered for precise measurement applications and is compatible with Brown & Sharpe Interapid 312 Dial Test Indicators, with the exception of models 74.111965 and 74.111958.
  • Ball style for accurate contact.
  • Features a 0.0800 in ball diameter.
  • Constructed from carbide for enhanced durability.
  • Designed for use with specific dial test indicators.
Catalog No. 50-005-36534 Supplier MSC Supplier No. 99225773
This test indicator contact point, with a 0.0800 in ball diameter, is crafted from durable carbide. It is engineered for precise measurement applications and is compatible with Brown & Sharpe Interapid 312 Dial Test Indicators, with the exception of models 74.111965 and 74.111958.
  • Ball style for accurate contact.
  • Features a 0.0800 in ball diameter.
  • Constructed from carbide for enhanced durability.
  • Designed for use with specific dial test indicators.