Learn More
MSC INTERAPID Test Indicator Contact Point, Ball, Carbide, 0.0310 in Ball Diameter, 1-3/8 in Length
Catalog No. 5000510423
Encompass
This contact point is made of carbide and features a 0.0310 in ball diameter with a length of 1-3/8 in. It is designed for use with Brown & Sharpe Interapid 312 Dial Test Indicators, excluding certain models.
  • Ball style for accurate measurements
  • 0.0310 in ball diameter for precise contact
  • 1-3/8 in contact point length for extended reach
  • Carbide construction ensures durability
  • Compatible with a range of dial test indicators
Catalog No. 50-005-10423 Supplier MSC Supplier No. 99036931
This contact point is made of carbide and features a 0.0310 in ball diameter with a length of 1-3/8 in. It is designed for use with Brown & Sharpe Interapid 312 Dial Test Indicators, excluding certain models.
  • Ball style for accurate measurements
  • 0.0310 in ball diameter for precise contact
  • 1-3/8 in contact point length for extended reach
  • Carbide construction ensures durability
  • Compatible with a range of dial test indicators